Learn More
Sino Biological Anti-Cortisol Binding Globulin/CBG Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10998MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SerpinA6 (rh SerpinA6; Catalog#10998-H08H; NP_001747.2; Met1-Val405). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8461