Learn More
Sino Biological Human SerpinA6 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10998RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SerpinA6 (rh SerpinA6; Catalog#10998-H08H; NP_001747.2; Met 1-Val 405). SerpinA6 specific IgG was purified by Human SerpinA6 affinity chromatography.
Catalog No. 50-237-2208