Learn More
Sino Biological Recombinant Human Cortisol Binding Globulin/CBG Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10998H27HB100
Encompass_Preferred
A DNA sequence encoding the human SERPINA6 (NP_001747.2) (Met1-Val405) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4988