Learn More
Sino Biological Human SerpinB1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 12588MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SerpinB1 / ELANH2 (rh SerpinB1 / ELANH2; Catalog#12588-H08B; P30740; Met 1-Pro 379). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3677