Learn More
Sino Biological Human SerpinB6 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 12837RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SerpinB6 (rh SerpinB6; Catalog#12837-H07B; AAB30320.1; Asp 2-Pro 376). SerpinB6 specific IgG was purified by Human SerpinB6 affinity chromatography.
Catalog No. 50-237-2899