Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SERPINC1 native Protein from human plasma

Catalog No. 89961544 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-544 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-961-544 Supplier Abnova™ Supplier No. P4952
Add to Cart
Add to Cart

Used for SDS-PAGE

The protein encoded by this gene is a plasma protease inhibitor and a member of the serpin superfamily. This protein inhibits thrombin as well as other activated serine proteases of the coagulation system, and it regulates the blood coagulation cascade. The protein includes two functional domains: the heparin binding-domain at the N-terminus of the mature protein, and the reactive site domain at the C-terminus. The inhibitory activity is enhanced by the presence of heparin. More than 120 mutations have been identified for this gene, many of which are known to cause antithrombin-III deficiency. [provided by RefSeq]

Specifications

Concentration 8.3 mg/mL
For Use With (Application) SDS-PAGE
Formulation In 0.05M Tris-HCl, 0.1M NaCl, pH 7.4
Gene ID (Entrez) 462
Molecular Weight (g/mol) 58kDa
Name SERPINC1 (Human) Native Protein
Preparation Method Native protein purified from human plasma
Purification Method Multi-step chromatography
Quantity 1 mg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias AT3/ATIII/MGC22579
Common Name SERPINC1
Gene Symbol SERPINC1
Species Human
Protein Tag None
Expression System Native protein purified from human plasma
Form Liquid
Purity or Quality Grade >98% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.