Learn More
Sino Biological Anti-PAI-1/Serpin E1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10296MM0120
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SerpinE1 / PAI-1 (rh SerpinE1 / PAI-1; Catalog#10296-H08H; NP_000593.1; Met1-Pro402). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0384