Learn More
Sino Biological Recombinant Rat Cripto Protein (His Tag)
Supplier:  Sino Biological 80135R08H50
Encompass_Preferred
A DNA sequence encoding the rat TDGF1 (XP_001056317.2) (Met1-Cys143) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6630