Learn More
Sino Biological Recombinant Cynomolgus, Rhesus NKG2A Protein (hFc Tag)
Supplier:  Sino Biological 90214C01H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus KLRC1 (NP_001271106.1 & Q9MZJ2) (Pro94-Leu233) was expressed with the Fc region of human IgG1 at the N-terminus. Cynomolgus and Rhesus KLRC1 sequences are identical.
Catalog No. 50-224-6629