Learn More
Sino Biological Anti-C1 inhibitor/C1-INH Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10995MM0620
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human C1 inhibitor / SerpinG1 (rh C1 inhibitor / SerpinG1; Catalog#10995-H08H; NP_000053.2; Met1-Ala500). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-9269