Learn More
Sino Biological Human SerpinG1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10995MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SerpinG1 (rh SerpinG1; Catalog#10995-H08H; NP_000053.2; Met 1-Ala 500). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3796