Learn More
Sino Biological Anti-C1 inhibitor/C1-INH Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10995RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SerpinG1 (rh SerpinG1; Catalog#10995-H08H; NP_000053.2; Met 1-Ala 500). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1989