Learn More
Sino Biological Recombinant Anti-Neuroserpin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11107R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human SerpinI1 (rh SerpinI1; Catalog#11107-H08H; NP_005016.1; Met 1-Leu 410).
Catalog No. 50-241-2245