Learn More
Sino Biological Human SerpinI1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11107MM08100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Neuroserpin / SerpinI1 (rh Neuroserpin / SerpinI1; Catalog#11107-H08H; NP_005016.1; Met1-Leu410). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-0807