Learn More
Sino Biological Human SerpinI1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11107RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SerpinI1 (rh SerpinI1; Catalog#11107-H08H; NP_005016.1; Met 1-Leu 410). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-3448