Learn More
Sino Biological Human SerpinI2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11008RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SerpinI2 (rh SerpinI2; Catalog#11008-H08H; NP_006208.1; Met 1-Leu 405). SerpinI2 specific IgG was purified by Human SerpinI2 affinity chromatography.
Catalog No. 50-237-2455