Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SERTAD3 Full-length ORF (NP_037500.2, 1 a.a. - 196 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89961573 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-573 25 μg
89-961-572 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-961-573 Supplier Abnova™ Supplier No. H00029946P01L
Only null left
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene was identified in a yeast two-hybrid assay employing the second subunit of human replication protein A as bait. It is localized to the nucleus and its expression is significantly higher in cancer cell lines compared to normal cell lines. This protein has also been shown to be a strong transcriptional co-activator. Alternative splicing has been observed at this locus and two variants, both encoding the same protein, have been identified. [provided by RefSeq]

Sequence: MVGGLKRKHSDLEEEEERWEWSPAGLQSYQQALLRISLDKVQRSLGPRAPSLRRHVLIHNTLQQLQAALRLAPAPALPPEPLFLGEEDFSLSATIGSILRELDTSMDGTEPPQNPVTPLGLQNEVPPQPDPVFLEALSSRYLGDSGLDDFFLDIDTSAVEKEPARAPPEPPHNLFCAPGSWEWNELDHIMEIILGS

Specifications

Accession Number NP_037500.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 29946
Molecular Weight (g/mol) 48.2kDa
Name SERTAD3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MVGGLKRKHSDLEEEEERWEWSPAGLQSYQQALLRISLDKVQRSLGPRAPSLRRHVLIHNTLQQLQAALRLAPAPALPPEPLFLGEEDFSLSATIGSILRELDTSMDGTEPPQNPVTPLGLQNEVPPQPDPVFLEALSSRYLGDSGLDDFFLDIDTSAVEKEPARAPPEPPHNLFCAPGSWEWNELDHIMEIILGS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias RBT1
Common Name SERTAD3
Gene Symbol SERTAD3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.