Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GeminiBio™ Human Serum AB (Off-the-Clot)
SDP

Catalog No. 11852106
Change view
Click to view available options
Sterilization Method:
Gamma-irradiated
Sequential-filtered, 100 nm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterilization Method
11-852-106 Sequential-filtered, 100 nm
11-852-107 Gamma-irradiated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For further manufacturing or research use only. Not for direct Therapeutic use.
Catalog No. 11-852-106 Supplier GeminiBio™ Supplier No. 100318750
Add to Cart
Add to Cart
For further manufacturing or research use only. Not for direct Therapeutic use.

Collected from healthy male AB donors and produced via natural coagulation with no anticoagulants or preservatives. Extensively tested to deliver consistent cell culture performance while supporting clinical and commercial use, traceability, and raw material control.

  • Human Serum AB, Off-the-Clot is designed for cell therapy programs that prioritize biological fidelity, defined sourcing, and regulatory readiness.
  • Produced through natural blood coagulation, this serum format preserves native composition while avoiding the use of anticoagulants or preservatives.
  • Extensively tested and sourced from a controlled donor pool, supporting clinical and commercial applications where consistency, traceability, and supply continuity matter.
  • Naturally coagulated (off-the-clot) serum format with no added anticoagulants or preservatives
  • Collected from healthy male donors of the AB serotype
  • Extensively tested to support clinical and commercial workflows
  • Defined donor pool (>50 donors) balancing traceability with supply continuity
  • Clear processing methodology supporting raw material control and consistency expectations

Suited for:

  • Clinical and commercial programs seeking a minimally processed Human Serum AB format
  • Applications prioritizing native serum characteristics
  • Teams requiring established testing, documentation, and long-term supply planning

Specifications

Form Frozen
Format Bag - BioProcess Container (BPC)
Product Type Human Serum
Quantity 100 mL
Sterility Sterile-filtered
Sterilization Method Sequential-filtered, 100 nm
Mycoplasma Detected No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.