Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Human Serum Albumin (HSA), lyophilized powder, 100 g
SDP
Catalog No. 502388576
Encompass_Preferred
PAN-Biotech Human Serum Albumin (HSA), lyophilized powder, 100 g. APPLICATION: Albumins are used to solubilize lipids and stabilize other proteins such as growth factors in tissue culture applications. Also used as blocking agents in Western blots or ELISA applications and as a protein standard. Albumin has a low molecular weight, is easily soluble in water and contains an excess of acidic amino acids. Used as serum substitute in serum-free, xeno-free cell culture. Medium component for cryopreservation and stem cell culture. Stabilizer protein for pharmaceutical and biotech products. Reduces unspecific binding in molecular biology assays. Manufactured per US and European Pharmacopeia specifications. Strict donor selection and screening. CAS NO.: 70024-90-7. COUNTRY OF ORIGIN: Germany. TYPE: Purified Protein, sterile. PURITY: HSA >98% pure according to agarose gel electrophoresis. FATTY ACID CONTENT: Not fatty acid free. PROTEASE CONTENT: Not protease free. STORAGE: 2-8°C.
Catalog No. 50-238-8576 Supplier ILEX LIFE SCIENCES LLC Supplier No. P06260100
PAN-Biotech Human Serum Albumin (HSA), lyophilized powder, 100 g. APPLICATION: Albumins are used to solubilize lipids and stabilize other proteins such as growth factors in tissue culture applications. Also used as blocking agents in Western blots or ELISA applications and as a protein standard. Albumin has a low molecular weight, is easily soluble in water and contains an excess of acidic amino acids. Used as serum substitute in serum-free, xeno-free cell culture. Medium component for cryopreservation and stem cell culture. Stabilizer protein for pharmaceutical and biotech products. Reduces unspecific binding in molecular biology assays. Manufactured per US and European Pharmacopeia specifications. Strict donor selection and screening. CAS NO.: 70024-90-7. COUNTRY OF ORIGIN: Germany. TYPE: Purified Protein, sterile. PURITY: HSA >98% pure according to agarose gel electrophoresis. FATTY ACID CONTENT: Not fatty acid free. PROTEASE CONTENT: Not protease free. STORAGE: 2-8°C.