Learn More
NEXPRING HEALTH Human Serum Albumin Solution, 12 x 5 mL
Catalog No. NC0622494
Encompass
Human Serum Albumin consists of HSA (100 mg/ml) from a therapeutic grade source material in a saline solution. This represents 10% total protein (weight/volume), in normal saline.
Catalog No. NC0622494 Supplier NEXPRING HEALTH Supplier No. 998812X5ML
Human Serum Albumin consists of HSA (100 mg/ml) from a therapeutic grade source material in a saline solution. This represents 10% total protein (weight/volume), in normal saline.