Learn More
Creative Biomart Human serum albumin, Albumin human
Catalog No. 502084126
Encompass_Preferred
The product is the main protein of human blood plasma. It binds to water, cations (such as Ca2+, Na+ and K+), fatty acids, hormones, bilirubin, thyroxine (T4) and pharmaceuticals (including barbiturates). The main function of albumin is to maintain the oncotic pressure of blood.
Catalog No. 50-208-4126 Supplier Creative Biomart Supplier No. THP0075
The product is the main protein of human blood plasma. It binds to water, cations (such as Ca2+, Na+ and K+), fatty acids, hormones, bilirubin, thyroxine (T4) and pharmaceuticals (including barbiturates). The main function of albumin is to maintain the oncotic pressure of blood.