Learn More
Medchemexpress LLC Human Serum Albumin-AF750 | 1 MG
SDP
Catalog No. 5000425671
Encompass_Preferred
Human Serum Albumin-AF750 is a biochemical reagent conjugated to fluorescein AF750. This fluorescent dye has an excitation wavelength of 752 nm and an emission wavelength of 780 nm. It serves as a versatile tool in various biological assays.
  • Used to label specific antigens, molecules, or cellular structures
  • Used to detect specific antigens, molecules, or cellular structures
  • Suitable for biochemical assay applications
  • Relevant for inflammation or immune system disease research
  • Relevant for blood or cardio-cerebrovascular disease research
Catalog No. 50-004-25671 Supplier Medchemexpress LLC Supplier No. HYNP2041MG
Add to Cart
Add to Cart
Human Serum Albumin-AF750 is a biochemical reagent conjugated to fluorescein AF750. This fluorescent dye has an excitation wavelength of 752 nm and an emission wavelength of 780 nm. It serves as a versatile tool in various biological assays.
  • Used to label specific antigens, molecules, or cellular structures
  • Used to detect specific antigens, molecules, or cellular structures
  • Suitable for biochemical assay applications
  • Relevant for inflammation or immune system disease research
  • Relevant for blood or cardio-cerebrovascular disease research