Learn More
Medchemexpress LLC Human Serum Albumin-AF750 | 100 UG
SDP
Catalog No. 5000425764
Encompass_Preferred
Human Serum Albumin-AF750 is a biochemical reagent conjugated to fluorescein AF750 (Ex=752 nM; Em=780 nm). It can be used to label or detect specific antigens, molecules, or cellular structures.
  • Conjugated to fluorescein AF750 (Ex=752 nM; Em=780 nm)
  • Labels specific antigens, molecules, or cellular structures
  • Detects specific antigens, molecules, or cellular structures
Catalog No. 50-004-25764 Supplier Medchemexpress LLC Supplier No. HYNP204100UG
Human Serum Albumin-AF750 is a biochemical reagent conjugated to fluorescein AF750 (Ex=752 nM; Em=780 nm). It can be used to label or detect specific antigens, molecules, or cellular structures.
  • Conjugated to fluorescein AF750 (Ex=752 nM; Em=780 nm)
  • Labels specific antigens, molecules, or cellular structures
  • Detects specific antigens, molecules, or cellular structures