Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Human Serum Albumin Antibody (YA984) | 100 UL
SDP

Catalog No. 5000442950
Encompass_Preferred

Human Serum Albumin Antibody (YA984) is a mouse-derived, non-conjugated IgG1 monoclonal antibody designed for research use only, targeting Human Serum Albumin. It binds water, ions, fatty acids, hormones, bilirubin, and drugs, primarily regulating the colloidal osmotic pressure of blood and acting as a major zinc, calcium, and magnesium transporter.

  • Mouse-derived IgG1 monoclonal antibody
  • Targeting Human Serum Albumin
  • For research use only
  • Supplied in PBS containing 50% glycerol, 0.5% BSA, and 0.02% sodium azide
  • Store at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles
  • Species reactivity is human
  • Applications include ELISA at 1:10000

Catalog No. 50-004-42950 Supplier Medchemexpress LLC Supplier No. HYP81281100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Human Serum Albumin Antibody (YA984) is a mouse-derived, non-conjugated IgG1 monoclonal antibody designed for research use only, targeting Human Serum Albumin. It binds water, ions, fatty acids, hormones, bilirubin, and drugs, primarily regulating the colloidal osmotic pressure of blood and acting as a major zinc, calcium, and magnesium transporter.

  • Mouse-derived IgG1 monoclonal antibody
  • Targeting Human Serum Albumin
  • For research use only
  • Supplied in PBS containing 50% glycerol, 0.5% BSA, and 0.02% sodium azide
  • Store at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles
  • Species reactivity is human
  • Applications include ELISA at 1:10000

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.