Learn More
Medchemexpress LLC Human serum albumin (cell culture grade, endotoxin<0.125 EU/mg) | 70024-90-7 | 99.6% | 25 MG
SDP
Catalog No. 5000425562
Encompass_Preferred
Human serum albumin (HSA) is the most abundant protein in plasma and a major determinant of plasma oncotic pressure. It possesses antioxidant, anticoagulant, anti-inflammatory, anti-platelet aggregation activities, and colloid osmotic action. This product is human serum albumin recombinantly expressed in an Escherichia coli expression system and is suitable for in vitro cell culture.
  • Blocks the inhibitory effect of GML on human T cells
  • Partially prevents LPS-induced oxidative stress
  • Contributes to blood pressure reduction
  • Suitable for in vitro cell culture
Catalog No. 50-004-25562 Supplier Medchemexpress LLC Supplier No. 25MGHYP1956A
Human serum albumin (HSA) is the most abundant protein in plasma and a major determinant of plasma oncotic pressure. It possesses antioxidant, anticoagulant, anti-inflammatory, anti-platelet aggregation activities, and colloid osmotic action. This product is human serum albumin recombinantly expressed in an Escherichia coli expression system and is suitable for in vitro cell culture.
  • Blocks the inhibitory effect of GML on human T cells
  • Partially prevents LPS-induced oxidative stress
  • Contributes to blood pressure reduction
  • Suitable for in vitro cell culture