Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Human Serum Albumin (HSA), lyophilized powder, 5g
SDP
Catalog No. NC2962008
Encompass_Preferred
Albumins are monomeric proteins found in body fluids and tissues. Human serum albumin (HSA) is the most abundant protein in human blood plasma with a molecular weight of about 66.5 kDa. The major biological function of albumin is to regulate the colloidal osmotic blood pressure. In addition, albumin acts as carrier protein for hormones, fatty acids and other compounds, and buffers pH. The reference range for albumin concentration in serum is approximately 35-50 g/L.
Catalog No. NC2962008 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0626005
Albumins are monomeric proteins found in body fluids and tissues. Human serum albumin (HSA) is the most abundant protein in human blood plasma with a molecular weight of about 66.5 kDa. The major biological function of albumin is to regulate the colloidal osmotic blood pressure. In addition, albumin acts as carrier protein for hormones, fatty acids and other compounds, and buffers pH. The reference range for albumin concentration in serum is approximately 35-50 g/L.