Learn More
Medchemexpress LLC Human serum albumin (HSA) | 70024-90-7 | 99.9% | 250 MG
SDP
Catalog No. 5000426218
Encompass_Preferred
Human serum albumin (HSA) is the most abundant protein in plasma and plays a crucial role in maintaining plasma oncotic pressure. It exhibits antioxidant, anticoagulant, anti-inflammatory, and anti-platelet aggregation activities, in addition to its colloid osmotic action. This recombinant Human Serum Albumin is expressed in a microbial expression system.
  • Plays a crucial role in maintaining plasma oncotic pressure.
  • Exhibits antioxidant activity.
  • Has anticoagulant properties.
  • Demonstrates anti-inflammatory effects.
  • Shows anti-platelet aggregation activities.
  • Provides protective function for T cells by blocking GML inhibitory effects.
  • Associated with cardiovascular diseases, preventing LPS-induced oxidative stress.
  • Contributes to blood pressure reduction by inhibiting NF-κB and peroxynitrite upregulation.
Catalog No. 50-004-26218 Supplier Medchemexpress LLC Supplier No. 250MGHYP1956
Human serum albumin (HSA) is the most abundant protein in plasma and plays a crucial role in maintaining plasma oncotic pressure. It exhibits antioxidant, anticoagulant, anti-inflammatory, and anti-platelet aggregation activities, in addition to its colloid osmotic action. This recombinant Human Serum Albumin is expressed in a microbial expression system.
  • Plays a crucial role in maintaining plasma oncotic pressure.
  • Exhibits antioxidant activity.
  • Has anticoagulant properties.
  • Demonstrates anti-inflammatory effects.
  • Shows anti-platelet aggregation activities.
  • Provides protective function for T cells by blocking GML inhibitory effects.
  • Associated with cardiovascular diseases, preventing LPS-induced oxidative stress.
  • Contributes to blood pressure reduction by inhibiting NF-κB and peroxynitrite upregulation.