Learn More
Medchemexpress LLC Human serum albumin (HSA) | 70024-90-7 | 99.9% | 5 G
SDP
Catalog No. 5000422920
Encompass_Preferred
Human serum albumin (HSA) is the most abundant protein in plasma and a major determinant of plasma oncotic pressure. It exhibits antioxidant, anticoagulant, anti-inflammatory, and anti-platelet aggregation activities, as well as colloid osmotic action. This recombinant Human Serum Albumin is expressed in a microbial expression system and can protect T cell function by inhibiting the effect of Glycerol Monolaurate (GML).
  • Most abundant protein in plasma
  • Major determinant of plasma oncotic pressure
  • Exhibits antioxidant, anticoagulant, anti-inflammatory, and anti-platelet aggregation activities
  • Provides colloid osmotic action
  • Protects T cell function
  • Partially prevents oxidative stress
  • Contributes to reduction of blood pressure
Catalog No. 50-004-22920 Supplier Medchemexpress LLC Supplier No. HYP19565G
Human serum albumin (HSA) is the most abundant protein in plasma and a major determinant of plasma oncotic pressure. It exhibits antioxidant, anticoagulant, anti-inflammatory, and anti-platelet aggregation activities, as well as colloid osmotic action. This recombinant Human Serum Albumin is expressed in a microbial expression system and can protect T cell function by inhibiting the effect of Glycerol Monolaurate (GML).
  • Most abundant protein in plasma
  • Major determinant of plasma oncotic pressure
  • Exhibits antioxidant, anticoagulant, anti-inflammatory, and anti-platelet aggregation activities
  • Provides colloid osmotic action
  • Protects T cell function
  • Partially prevents oxidative stress
  • Contributes to reduction of blood pressure