Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GeminiBio™ Human Serum Albumin (HSA) Powder - Low Endotoxin
SDP

Catalog No. 11852102 Shop All Gemini Bio Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-852-102 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For further manufacturing or research use only. Not for direct Therapeutic use.
Catalog No. 11-852-102 Supplier GeminiBio™ Supplier No. 800126P5G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For further manufacturing or research use only. Not for direct Therapeutic use.

Designed for research and diagnostic applications.

  • High-purity Human Albumin produced by cold-alcohol fractionation from pooled human plasma and lyophilized.
  • MW ∼66 kDa.
  • Not for clinical or therapeutic use.
  • Endotoxin ≤2 EU/mg.

Specifications

Quantity 5 g
Species Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.