Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GeminiBio™ Human Serum Albumin (HSA) Powder
SDP

Catalog No. 11852105
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-852-105 25 g
11-852-101 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For further manufacturing or research use only. Not for direct Therapeutic use.
Catalog No. 11-852-105 Supplier GeminiBio™ Supplier No. 800125P25G
Only null left
Add to Cart
Add to Cart
For further manufacturing or research use only. Not for direct Therapeutic use.

Designed for research and diagnostic applications.

  • High-purity Human Albumin produced by cold-alcohol fractionation from pooled human plasma and lyophilized for stability and ease of use.
  • MW ∼66 kDa.
  • Not for clinical or therapeutic use.

Specifications

Form Lyophilized
For Use With (Application) Cell Culture Supplement, Protein Stabilizer, Carrier Protein, Assay Reagent Component, Diagnostic Manufacturing, Research Use
Quantity 25 g
Species Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.