Learn More
Description
- Contains USP-required sodium caprylate and tryptophanate preservatives.
- For research and diagnostic use only.
- Ships frozen.
Specifications
Specifications
| Concentration | 25% |
| Form | Liquid |
| Product Type | Human Serum Albumin (HSA) |
| For Use With (Application) | Cell Culture Supplement, Protein Stabilizer, Carrier Protein, Assay Reagent Component, Diagnostic Manufacturing, Research Use |
| Quantity | 50 mL |
| Species | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
