Learn More
American Research Products Inc Human Serum amyloid A 4 protei
Catalog No. 5000327996
Encompass
Human Serum amyloid A-4 protein (SAA4) ELISA kit from CUSABIO Detection Range 15 6 ng/ml-1000 ng/ml Sensitivity 3 9 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27996 Supplier American Research Products Inc Supplier No. CSBEL020659HU
Human Serum amyloid A-4 protein (SAA4) ELISA kit from CUSABIO Detection Range 15 6 ng/ml-1000 ng/ml Sensitivity 3 9 ng/ml Sample serum plasma tissue homogenates Method Sandwich