Learn More
Biomatik Corporation Human Serum Amyloid A (SAA) CLIA Kit
Supplier:  Biomatik Corporation EKU09895
Encompass_Preferred
Human Serum Amyloid A (SAA) CLIA Kit for detecting SAA in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 54.9-40,000pg/ml. Sensitivity: 22.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P0DJI8. Gene ID: 6288. Target Synonyms: SA-A; SAA1; PIG4; TP53I4; Amyloid protein A; Amyloid fibril protein AA; Serum amyloid A-1 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5194