Learn More
Biomatik Corporation Human Serum Amyloid P Component (SAP) CLIA Kit
Supplier:  Biomatik Corporation EKU09774
Encompass_Preferred
Human Serum Amyloid P Component (SAP) CLIA Kit for detecting SAP in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.55-400ng/ml. Sensitivity: 0.23ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P02743. Gene ID: 325. Target Synonyms: APCS; PTX2; Pentaxin-Related; 9.5S Alpha-1-Glycoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7348