Learn More
Sigma Aldrich Fine Chemicals Biosciences Human serum serum proteins
Catalog No. 502904632
Encompass
ERM-DA470k/IfCC replaces the former product ERM-DA470. The new reference material is certified for 12 serum proteins. The material is the lyophilised form of processed serum and is certified for A2M AAG AAT ALB C3C C4 HPT IgA IgG IgM TRf and TTR.
Catalog No. 50-290-4632 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. ERMDA470KIFCC1VL
ERM-DA470k/IfCC replaces the former product ERM-DA470. The new reference material is certified for 12 serum proteins. The material is the lyophilised form of processed serum and is certified for A2M AAG AAT ALB C3C C4 HPT IgA IgG IgM TRf and TTR.