Learn More
Biomatik Corporation Human Serum/Glucocorticoid Regulated Kinase 1 (SGK1) ELISA Kit
Supplier:  Biomatik Corporation EKU12183
Encompass_Preferred
Human Serum/Glucocorticoid Regulated Kinase 1 (SGK1) ELISA Kit for detecting SGK1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.147ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O00141. Gene ID: 6446. Target Synonyms: Serine/threonine-protein kinase Sgk1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4584