Learn More
ACROBiosystems HEK293/Human SEZ6 Stable Cell Line
Catalog No. 502719731
Encompass_Preferred
May play a role in cell-cell recognition and in neuronal membrane signaling. Seems to be important for the achievement of the necessary balance between dendrite elongation and branching during the elaboration of a complex dendritic arbor. Involved in the development of appropriate excitatory synaptic connectivity
Catalog No. 50-271-9731 Supplier ACROBiosystems Supplier No. CHEKATP1832VIALS
May play a role in cell-cell recognition and in neuronal membrane signaling. Seems to be important for the achievement of the necessary balance between dendrite elongation and branching during the elaboration of a complex dendritic arbor. Involved in the development of appropriate excitatory synaptic connectivity