Learn More
Sino Biological Anti-14-3-3 sigma Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10838MM02P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human 14-3-3 sigma / Stratifin / YWHAS (rh 14-3-3 sigma / Stratifin / YWHAS; Catalog#10838-HNCE; NP_006133.1; Met1-Ser248) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-255-5138