Learn More
Sino Biological Anti-14-3-3 sigma Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10838RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human 14-3-3 sigma / Stratifin / YWHAS / SFN (rh 14-3-3 sigma / Stratifin / YWHAS / SFN; Catalog#10838-HNCE; NP_006133.1; Met 1-Ser 248). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-5136