Learn More
Sino Biological Anti-SFRP1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10680T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SFRP1 (rh SFRP1; Catalog#10680-H08H; NP_003003.3; Met1-Lys314). SFRP1 specific IgG was purified by Human SFRP1 affinity chromatography.
Catalog No. 50-240-8292