Learn More
Sino Biological Anti-SP-D/SFTPD Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11041RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SFTPD / SP-D / SFTP4 (rh SFTPD / SP-D / SFTP4; Catalog#11041-H08H; NP_003010.4; Met1-Phe375). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2301