Learn More
Sino Biological Recombinant Human SAP/SH2D1A Protein (His Tag)
Catalog No. 502993223
Encompass_Preferred
A DNA sequence encoding the Human SH2D1A (NP_002342.1) (Met1-Pro128) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-299-3223 Supplier Sino Biological Supplier No. 11149H07E1100
A DNA sequence encoding the Human SH2D1A (NP_002342.1) (Met1-Pro128) was expressed with a polyhistidine tag at the N-terminus.