Learn More
Biomatik Corporation Human SH3 Domain Binding Glutamic Acid Rich Protein Like Protein (SH3BGRL) ELISA Kit
Supplier:  Biomatik Corporation EKU07319
Encompass_Preferred
Human SH3 Domain Binding Glutamic Acid Rich Protein Like Protein (SH3BGRL) ELISA Kit for detecting SH3BGRL in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75368. Gene ID: 6451. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4891