Learn More
BIOHIPPO Human SHH Microsample
SDP
Catalog No. 5000314013
Encompass
Human SHH Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SHH in tissue homogenates cell lysates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 28 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human SHH Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human SHH Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human SHH biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14013 Supplier BIOHIPPO Supplier No. BHE15212460
Human SHH Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SHH in tissue homogenates cell lysates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 28 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human SHH Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human SHH Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human SHH biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color