Learn More
BIOHIPPO Human sHLA-G Microsample
SDP
Catalog No. 5000315072
Encompass
Human sHLA-G Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of sHLA-G in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 066 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human sHLA-G Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human sHLA-G Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human sHLA-G biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15072 Supplier BIOHIPPO Supplier No. BHE15210544
Human sHLA-G Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of sHLA-G in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 066 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human sHLA-G Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human sHLA-G Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human sHLA-G biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color