Learn More
Biomatik Corporation Human Short Palate, Lung And Nasal Epithelium Carcinoma Associated Protein 2 (SPLUNC2) ELISA Kit
Supplier:  Biomatik Corporation EKU07322
Encompass_Preferred
Human Short Palate, Lung And Nasal Epithelium Carcinoma Associated Protein 2 (SPLUNC2) ELISA Kit for detecting SPLUNC2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96DR5. Gene ID: 140683. Target Synonyms: BPIFA2; PSP; C20orf70; BPI fold-containing family A member 2; Parotid secretory protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8761