Learn More
Sino Biological Anti-SIAE Antibody, Mouse Monoclonal
Supplier:  Sino Biological 13310MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SIAE/sialate O-acetylesterase (rh SIAE/sialate O-acetylesterase; Catalog#13310-H08B; Q9HAT2-1; Met1-Lys523). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1422