Learn More
Biomatik Corporation Human Sialic Acid Acetylesterase (SIAE) ELISA Kit
Supplier:  Biomatik Corporation EKU07324
Encompass_Preferred
Human Sialic Acid Acetylesterase (SIAE) ELISA Kit for detecting SIAE in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9HAT2. Gene ID: 54414. Target Synonyms: YSG2; CSE-C; LSE; Sialic Acid-Specific Acetylesterase II; Sialic acid-specific 9-O-acetylesterase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8759