Learn More
Biomatik Corporation Human Sialic Acid Binding Ig Like Lectin 10 (SIGLEC10) ELISA Kit
Supplier:  Biomatik Corporation EKU07326
Encompass_Preferred
Human Sialic Acid Binding Ig Like Lectin 10 (SIGLEC10) ELISA Kit for detecting SIGLEC10 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.12-5000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96LC7. Gene ID: 89790. Target Synonyms: PRO940; SLG2; Sialic Acid Binding Ig-Like Lectin 10 Ig-Like Lectin 7; Siglec-Like Gene 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4882