Learn More
Biomatik Corporation Human Sialic Acid Binding Ig Like Lectin 15 (SIGLEC15) ELISA Kit
Supplier:  Biomatik Corporation EKU12262
Encompass_Preferred
Human Sialic Acid Binding Ig Like Lectin 15 (SIGLEC15) ELISA Kit for detecting SIGLEC15 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6ZMC9. Gene ID: 284266. Target Synonyms: CD33L3; CD33 antigen-like 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5130